Gene Transcripts

Gene Symbol
NUDT22
Entrez Gene ID
84304
Gene Name
nudix hydrolase 22
Gene Aliases
Chromosome Location
Chr. 11: 64226258 - 64230016 on Build GRCh38
UniGene ID
Hs.656074
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128612.2 NP_001122084.1 3, 4 630
NM_032344.3 NP_115720.1 3, 4 744
GenBank mRNA BC006129.1 - 3, 4 728
BG705568.1 - 3, 4 635

View Full Product Details