Gene Transcripts

Gene Symbol
APOPT1
Entrez Gene ID
84334
Gene Name
apoptogenic 1, mitochondrial
Gene Aliases
APOP, APOP1, C14orf153
Chromosome Location
Chr. 14: 103562957 - 103592187 on Build GRCh38
UniGene ID
Hs.598441
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001302652.1 NP_001289581.1 3, 4 438
NM_032374.4 NP_115750.2 3, 4 438
NR_126431.1 4, 5 603
GenBank mRNA AK223092.1 - 3, 4 409
AK223103.1 - 3, 4 409
BC007412.2 - 3, 4 401 2989951
BF528062.1 - 4, 5 505
BP368962.1 - 3, 4 432
BU181154.1 - 433
CB136383.1 - 3, 4 433

View Full Product Details