Gene Transcripts

Gene Symbol
FAM213A
Entrez Gene ID
84293
Gene Name
family with sequence similarity 213 member A
Gene Aliases
C10orf58, PAMM
Chromosome Location
Chr. 10: 80407829 - 80437115 on Build GRCh38
UniGene ID
Hs.500333
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243778.1 NP_001230707.1 4 410
NM_001243779.1 NP_001230708.1 4 480
NM_001243780.1 NP_001230709.1 4 976
NM_001243781.1 NP_001230710.1 4 493
NM_001243782.1 NP_001230711.1 4 393
NM_032333.4 NP_115709.3 4 510
XM_011540265.2 XP_011538567.1 4 813
XM_011540266.2 XP_011538568.1 4 1216
XM_017016775.1 XP_016872264.1 4 497
GenBank mRNA AF305824.1 - 2 166
AK074643.1 - 4 455
AK075447.1 - 4 485
AK225387.1 - 4 384
AK315440.1 - 4 404
AY359059.1 - 4 403
BC005871.2 - 4 372 3010078
BE742158.1 - 4 480
CN396006.1 - 4 362

View Full Product Details