Gene Transcripts

Gene Symbol
MGARP
Entrez Gene ID
84709
Gene Name
mitochondria localized glutamic acid rich protein
Gene Aliases
C4orf49, CESP-1, HUMMR, OSAP
Chromosome Location
Chr. 4: 139266163 - 139280338 on Build GRCh38
UniGene ID
Hs.710036
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032623.3 NP_116012.2 4 542
GenBank mRNA AF329088.1 - 4 429
AF484960.1 - 4 542
BC104173.1 - 4 428 40030666
BC104174.1 - 4 428 40030667

View Full Product Details