Gene Transcripts

Gene Symbol
CCDC62
Entrez Gene ID
84660
Gene Name
coiled-coil domain containing 62
Gene Aliases
CT109, ERAP75, TSP-NY
Chromosome Location
Chr. 12: 122773621 - 122827385 on Build GRCh38
UniGene ID
Hs.592008
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201435.4 NP_958843.2 7 958
NR_027918.2 7 958
XM_006719643.1 XP_006719706.1 7 866
XM_006719644.1 XP_006719707.1 7 866
XM_011538856.1 XP_011537158.1 7 863
XM_011538857.1 XP_011537159.1 7 866
XM_011538859.1 XP_011537161.1 4 329
XM_017020033.1 XP_016875522.1 6 1744
GenBank mRNA AK097663.1 - 7 882
AK124633.1 - 3 1282
AK292466.1 - 7 866
AY009105.1 - 3 274
AY254201.1 - 7 939
BC060796.1 - 7 941 5284906

View Full Product Details