Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001198696.1 | NP_001185625.1 | 5, 6 | 732 | |
NM_032797.5 | NP_116186.1 | 5, 6 | 729 | ||
GenBank mRNA | AF337957.1 | - | 4, 5 | 533 | |
AF506757.1 | - | 5, 6 | 729 | ||
AK027403.1 | - | 5, 6 | 608 | ||
AK127353.1 | - | 5, 6 | 2484 | ||
BC006121.1 | - | 4, 5 | 537 | 3506309 | |
BC023601.2 | - | 5, 6 | 716 | 4548458 | |
BX537621.1 | - | 3, 4 | 329 |