Gene Transcripts

Gene Symbol
XAGE2
Entrez Gene ID
9502
Gene Name
X antigen family member 2
Gene Aliases
CT12.2, GAGED3, XAGE-2, XAGE2B
Chromosome Location
Chr. X: 52368996 - 52375683 on Build GRCh38
UniGene ID
Hs.409726
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130777.2 NP_570133.1 4 434
XM_011530836.2 XP_011529138.1 4 462
GenBank mRNA AJ318880.1 - 4 409
BC009232.2 - 4 414 3960352
BC066311.1 - 4 416 30330532

View Full Product Details