Gene Transcripts

Gene Symbol
SLC22A9
Entrez Gene ID
114571
Gene Name
solute carrier family 22 member 9
Gene Aliases
HOAT4, OAT4, OAT7, UST3H, ust3
Chromosome Location
Chr. 11: 63368871 - 63410878 on Build GRCh38
UniGene ID
Hs.502772
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080866.2 NP_543142.2 3 794
XM_017017159.1 XP_016872648.1 3 1712
GenBank mRNA AB062418.1 - 3 627
AB074812.1 - 3 775
AF440402.1 - 3 762
AJ295270.1 - 3 586
AK074246.1 - 3 794
BC126288.1 - 3 676 8992004

View Full Product Details