Gene Transcripts

Gene Symbol
MTG1
Entrez Gene ID
92170
Gene Name
mitochondrial ribosome associated GTPase 1
Gene Aliases
GTP, GTPBP7
Chromosome Location
Chr. 10: 133394117 - 133420670 on Build GRCh38
UniGene ID
Hs.501578
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138384.2 NP_612393.2 4 416
GenBank mRNA AK074976.1 - 4 337
AK125012.1 - 5 703
AK292515.1 - 4 258
AK293988.1 - 4 324
AK308997.1 - 4 365
BC000920.2 - 2 125
BC004409.1 - 4 375
BC026039.1 - 4 392 5444745
BC026725.1 - 4 295
BC035721.1 - 4 320 5588933
DB136242.1 - 4 416

View Full Product Details