Gene Transcripts

Gene Symbol
ORMDL3
Entrez Gene ID
94103
Gene Name
ORMDL sphingolipid biosynthesis regulator 3
Gene Aliases
Chromosome Location
Chr. 17: 39921041 - 39927804 on Build GRCh38
UniGene ID
Hs.514151
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320801.1 NP_001307730.1 6 2794
NM_001320802.1 NP_001307731.1 4 1632
NM_001320803.1 NP_001307732.1 4 1346
NM_139280.3 NP_644809.1 4 1637
XM_017025345.1 XP_016880834.1 4 1490
XM_017025346.1 XP_016880835.1 4 1403
XM_017025347.1 XP_016880836.1 4 1583
XM_017025348.1 XP_016880837.1 4 1329
GenBank mRNA AF151043.1 - 1 76
AF373101.1 - 4 1373
AK074811.1 - 4 1434
AX775843.1 - 4 1391
BC017087.1 - 4 1403 3850211
BC018837.1 - 1 107
BC071833.1 - 4 1382 6340437

View Full Product Details