Gene Transcripts

Gene Symbol
STRA13
Entrez Gene ID
201254
Gene Name
stimulated by retinoic acid 13
Gene Aliases
CENP-X, CENPX, D9, FAAP10, MHF2
Chromosome Location
Chr. 17: 82018703 - 82022930 on Build GRCh38
UniGene ID
Hs.37616
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271007.1 NP_001257936.1 2, 3 128
NM_144998.3 NP_659435.2 2, 3 128
XM_017024329.1 XP_016879818.1 2, 3 149
GenBank mRNA BC009571.1 - 2, 3 97
BC011610.1 - 2, 3 79
BI837753.1 - 2, 3 96
U95006.1 - 2, 3 83
U95007.1 - 2, 3 83

View Full Product Details