Gene Transcripts

Gene Symbol
C9orf72
Entrez Gene ID
203228
Gene Name
chromosome 9 open reading frame 72
Gene Aliases
ALSFTD, FTDALS, FTDALS1
Chromosome Location
Chr. 9: 27546546 - 27573866 on Build GRCh38
UniGene ID
Hs.493639
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256054.2 NP_001242983.1 2 335
NM_018325.4 NP_060795.1 2 240
NM_145005.6 NP_659442.2 2 257
GenBank mRNA AK057806.1 - 2 235
AK291425.1 - 2 230
AL832229.1 - 1 197
AL832467.1 - 2 196
BC020851.2 - 1 177
BC068445.1 - 1 189 5298741
BG718065.1 - 2 246
BG722125.1 - 2 233
BQ068108.1 - 346
JN681271.1 - 2 313

View Full Product Details