Gene Transcripts

Gene Symbol
MBNL2
Entrez Gene ID
10150
Gene Name
muscleblind like splicing regulator 2
Gene Aliases
MBLL, MBLL39, PRO2032
Chromosome Location
Chr. 13: 97141780 - 97394120 on Build GRCh38
UniGene ID
Hs.657347
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001306070.1 NP_001292999.1 5 1538
NM_144778.3 NP_659002.1 5 1531
NM_207304.2 NP_997187.1 5 1531
XM_005254018.2 XP_005254075.1 5 1816
XM_005254019.2 XP_005254076.1 5 1816
XM_005254021.2 XP_005254078.1 5 1816
XM_005254022.2 XP_005254079.1 5 1816
XM_006719912.2 XP_006719975.1 5 1816
XM_011521045.2 XP_011519347.1 5 1423
XM_017020308.1 XP_016875797.1 5 1423
XM_017020309.1 XP_016875798.1 5 1407
XM_017020310.1 XP_016875799.1 5 1369
XM_017020311.1 XP_016875800.1 5 1407
GenBank mRNA AF061261.1 - 5 1473
AF491866.1 - 5 1472
AK291727.1 - 5 1478
AK315844.1 - 5 1454
AY101770.1 - 4 766
BC020418.1 - 5 1355
BC020430.1 - 4 984
BC032426.1 - 5 1462
BC104038.1 - 5 1459 40026585
BC104039.1 - 5 1459 40026586
BC104040.1 - 5 1459 40026588
CR749802.1 - 6 1490

View Full Product Details