Gene Transcripts

Gene Symbol
SLFN5
Entrez Gene ID
162394
Gene Name
schlafen family member 5
Gene Aliases
Chromosome Location
Chr. 17: 35243026 - 35273650 on Build GRCh38
UniGene ID
Hs.709347
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144975.3 NP_659412.3 2 621
XM_005257934.3 XP_005257991.1 2 662
GenBank mRNA AK054668.1 - 2 616
AK303299.1 - 2 616
AK303632.1 - 2 618
BC021238.1 - 2 599
BC125200.1 - 1 557 40123369
BC125201.1 - 1 557 40123372
BX647942.1 - 2 618

View Full Product Details