Gene Transcripts

Gene Symbol
KIF6
Entrez Gene ID
221458
Gene Name
kinesin family member 6
Gene Aliases
C6orf102, dJ1043E3.1, dJ137F1.4, dJ188D3.1
Chromosome Location
Chr. 6: 39335100 - 39725405 on Build GRCh38
UniGene ID
Hs.588202
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289021.1 NP_001275950.1 15 1756
NM_001289024.1 NP_001275953.1 3 397
NM_145027.4 NP_659464.3 16 1924
XM_005248904.4 XP_005248961.1 16 1924
XM_011514357.2 XP_011512659.1 16 1849
XM_011514358.2 XP_011512660.1 16 1849
XM_011514359.2 XP_011512661.1 16 1849
XM_017010427.1 XP_016865916.1 15 1814
XM_017010428.1 XP_016865917.1 11 1274
GenBank mRNA AK123217.1 - 3 397
AK131471.1 - 16 1924
AL832634.2 - 3 160
BC022074.2 - 3 338
BC098418.1 - 3 356
BC117307.1 - 15 1686 40125858
BC143445.1 - 15 1686 9051953
BX649045.1 - 13 1504
DA167995.1 - 3 397

View Full Product Details