Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001206774.1 | NP_001193703.1 | 2 | 283 | |
NM_012086.4 | NP_036218.1 | 2 | 283 | ||
XM_005246965.4 | XP_005247022.1 | 2 | 198 | ||
GenBank mRNA | AF133123.1 | - | 2 | 219 | |
AF465407.1 | - | 2 | 174 | ||
AK025560.1 | - | 2 | 197 | ||
AK314718.1 | - | 2 | 224 | ||
BC043347.1 | - | 2 | 138 | 5498609 | |
DB078021.1 | - | 2 | 283 |