Gene Transcripts

Gene Symbol
GORAB
Entrez Gene ID
92344
Gene Name
golgin, RAB6 interacting
Gene Aliases
GO, NTKLBP1, SCYL1BP1
Chromosome Location
Chr. 1: 170532122 - 170553834 on Build GRCh38
UniGene ID
Hs.183702
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146039.1 NP_001139511.1 1 140
NM_001320252.1 NP_001307181.1 1 140
NM_152281.2 NP_689494.2 1 140
NR_027397.1 1 140
XM_006711628.3 XP_006711691.1 1 111
XM_011510150.2 XP_011508452.1 1 126
GenBank mRNA AK021814.1 - 1 133
AK057661.1 - 1 90
AK308339.1 - 1 140
BC047476.1 - 1 111 5296051
BC064945.1 - 1 131 6019822

View Full Product Details