Gene Transcripts

Gene Symbol
TEX26
Entrez Gene ID
122046
Gene Name
testis expressed 26
Gene Aliases
C13orf26
Chromosome Location
Chr. 13: 30932633 - 30975502 on Build GRCh38
UniGene ID
Hs.124463
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152325.1 NP_689538.1 2 128
XM_011534919.2 XP_011533221.1 2 188
XM_011534921.2 XP_011533223.1 2 192
XM_011534924.2 XP_011533226.1 2 192
XM_011534925.2 XP_011533227.1 2 192
XM_011534926.2 XP_011533228.1 2 192
XM_011534927.2 XP_011533229.1 2 192
GenBank mRNA BC030277.1 - 2 128

View Full Product Details