Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145255.3 | NP_660298.2 | 3 | 359 | |
NM_148887.2 | NP_683685.1 | 4 | 457 | ||
NR_037575.1 | 3 | 508 | |||
GenBank mRNA | AK127167.1 | - | 3 | 1234 | |
AK293624.1 | - | 3 | 497 | ||
AL558270.3 | - | 4 | 422 | ||
AL701268.1 | - | 3 | 356 | ||
BC015904.1 | - | 3 | 330 | 3919892 | |
BC039852.1 | - | 3 | 338 | ||
BC052601.1 | - | 3 | 348 | 6085551 | |
DA137043.1 | - | 3 | 359 |