Gene Transcripts

Gene Symbol
LSM12
Entrez Gene ID
124801
Gene Name
LSM12 homolog
Gene Aliases
PNAS-135
Chromosome Location
Chr. 17: 44034635 - 44067619 on Build GRCh38
UniGene ID
Hs.355570
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152344.3 NP_689557.1 6 1927
GenBank mRNA AK055218.1 - 5 1656
AL832076.1 - 1 1615
BC016791.2 - 1 1421
BC044587.1 - 5 1657 4794614
DQ786277.1 - 1 40

View Full Product Details