Gene Transcripts
- Gene Symbol
- CNIH3
- Entrez Gene ID
- 149111
- Gene Name
- cornichon family AMPA receptor auxiliary protein 3
- Gene Aliases
- CNIH-3
- Chromosome Location
- Chr. 1: 224434640 - 224740549 on Build GRCh38
- UniGene ID
- Hs.731723
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001322302.1 |
NP_001309231.1
|
3, 4 |
438 |
|
NM_001322303.1 |
NP_001309232.1
|
2, 3 |
372 |
|
NM_001322304.1 |
NP_001309233.1
|
3, 4 |
516 |
|
NM_152495.1 |
NP_689708.1
|
2, 3 |
858 |
|
NR_136287.1 |
|
2, 3 |
858 |
|
NR_136288.1 |
|
3, 4 |
368 |
|
NR_136289.1 |
|
4, 5 |
504 |
|
NR_136290.1 |
|
3, 4 |
438 |
|
NR_136291.1 |
|
4, 5 |
574 |
|
NR_136292.1 |
|
3, 4 |
508 |
|
NR_136293.1 |
|
6, 7 |
744 |
|
NR_136294.1 |
|
5, 6 |
678 |
|
NR_136295.1 |
|
4, 5 |
549 |
|
NR_136296.1 |
|
4, 5 |
574 |
|
NR_136297.1 |
|
3, 4 |
438 |
|
GenBank mRNA |
AF070524.1 |
- |
1, 2 |
64 |
|
AK096312.1 |
- |
2, 3 |
858 |
|
BC022780.1 |
- |
2, 3 |
346 |
3894832 |
View Full Product Details