Gene Transcripts
- Gene Symbol
- ALS2CR11
- Entrez Gene ID
- 151254
- Gene Name
- amyotrophic lateral sclerosis 2 chromosome region candidate 11
- Gene Aliases
- Chromosome Location
- Chr. 2: 201487421 - 201619182 on Build GRCh38
- UniGene ID
- Hs.335788
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001168216.1 |
NP_001161688.1
|
5, 6 |
690 |
|
NM_001168217.1 |
NP_001161689.1
|
5, 6 |
690 |
|
NM_001168221.1 |
NP_001161693.1
|
5, 6 |
690 |
|
NM_152525.5 |
NP_689738.3
|
5, 6 |
690 |
|
XM_006712331.3 |
XP_006712394.1
|
5, 6 |
690 |
|
XM_006712332.3 |
XP_006712395.1
|
5, 6 |
690 |
|
XM_006712333.3 |
XP_006712396.1
|
5, 6 |
690 |
|
XM_006712334.3 |
XP_006712397.1
|
5, 6 |
690 |
|
XM_006712335.3 |
XP_006712398.1
|
5, 6 |
690 |
|
XM_006712336.3 |
XP_006712399.1
|
5, 6 |
690 |
|
XM_011510736.2 |
XP_011509038.1
|
5, 6 |
690 |
|
GenBank mRNA |
AK058080.1 |
- |
5, 6 |
662 |
|
AL833429.1 |
- |
5, 6 |
687 |
|
BC030659.2 |
- |
5, 6 |
683 |
4838801 |
BG773299.1 |
- |
5, 6 |
686 |
|
BX648732.1 |
- |
5, 6 |
683 |
|
View Full Product Details