Gene Transcripts
- Gene Symbol
- EXD1
- Entrez Gene ID
- 161829
- Gene Name
- exonuclease 3'-5' domain containing 1
- Gene Aliases
- EXDL1
- Chromosome Location
- Chr. 15: 41182728 - 41231074 on Build GRCh38
- UniGene ID
- Hs.307999
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001286441.1 |
NP_001273370.1
|
5, 6 |
730 |
|
NM_152596.3 |
NP_689809.2
|
3, 4 |
475 |
|
XM_006720415.3 |
XP_006720478.1
|
4, 5 |
647 |
|
XM_011521298.2 |
XP_011519600.1
|
6, 7 |
960 |
|
XM_011521299.2 |
XP_011519601.1
|
6, 7 |
691 |
|
XM_011521300.2 |
XP_011519602.1
|
3, 4 |
219 |
|
XM_011521301.2 |
XP_011519603.1
|
5, 6 |
516 |
|
XM_017021970.1 |
XP_016877459.1
|
3, 4 |
219 |
|
XM_017021971.1 |
XP_016877460.1
|
3, 4 |
217 |
|
GenBank mRNA |
AK292524.1 |
- |
3, 4 |
381 |
|
AK302864.1 |
- |
5, 6 |
664 |
|
BC030628.2 |
- |
3, 4 |
415 |
4827307 |
View Full Product Details