Gene Transcripts

Gene Symbol
CCDC185
Entrez Gene ID
164127
Gene Name
coiled-coil domain containing 185
Gene Aliases
C1orf65
Chromosome Location
Chr. 1: 223393373 - 223395470 on Build GRCh38
UniGene ID
Hs.192090
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152610.2 NP_689823.2 1 1767
GenBank mRNA AK093047.1 - 1 1723
BC030026.1 - 1 1716 4822648
BC031066.1 - 1 1714 5272765

View Full Product Details