Gene Transcripts

Gene Symbol
OIT3
Entrez Gene ID
170392
Gene Name
oncoprotein induced transcript 3
Gene Aliases
LZP
Chromosome Location
Chr. 10: 72893556 - 72933036 on Build GRCh38
UniGene ID
Hs.8366
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152635.2 NP_689848.1 5 927
NR_130125.1 5 927
GenBank mRNA AK096435.1 - 5 902
AY013707.2 - 5 758
AY358339.1 - 5 880
BC126440.1 - 5 899 8992156

View Full Product Details