Gene Transcripts
- Gene Symbol
- SLC44A5
- Entrez Gene ID
- 204962
- Gene Name
- solute carrier family 44 member 5
- Gene Aliases
- CTL5
- Chromosome Location
- Chr. 1: 75202131 - 75724261 on Build GRCh38
- UniGene ID
- Hs.654821
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001130058.1 |
NP_001123530.1
|
4, 5 |
253 |
|
NM_001320283.1 |
NP_001307212.1
|
2, 3 |
214 |
|
NM_152697.4 |
NP_689910.2
|
4, 5 |
253 |
|
XM_006710445.3 |
XP_006710508.1
|
2, 3 |
140 |
|
XM_017000607.1 |
XP_016856096.1
|
3, 4 |
279 |
|
XM_017000608.1 |
XP_016856097.1
|
3, 4 |
278 |
|
XM_017000609.1 |
XP_016856098.1
|
6, 7 |
1597 |
|
XM_017000610.1 |
XP_016856099.1
|
6, 7 |
675 |
|
XM_017000611.1 |
XP_016856100.1
|
4, 5 |
360 |
|
GenBank mRNA |
AK093170.1 |
- |
4, 5 |
255 |
|
AK296924.1 |
- |
3, 4 |
292 |
|
AK296941.1 |
- |
3, 4 |
198 |
|
AK299573.1 |
- |
2, 3 |
214 |
|
BC028743.2 |
- |
4, 5 |
226 |
4828797 |
BG718149.1 |
- |
4, 5 |
258 |
|
DB450780.1 |
- |
4, 5 |
254 |
|
DC374918.1 |
- |
2, 3 |
214 |
|
View Full Product Details