Gene Transcripts

Gene Symbol
LINC00521
Entrez Gene ID
256369
Gene Name
long intergenic non-protein coding RNA 521
Gene Aliases
C14orf48, c14_5713
Chromosome Location
Chr. 14: 93997270 - 94011695 on Build GRCh38
UniGene ID
Hs.143845
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024183.1 2, 3 516
NR_024184.1 2, 3 516
GenBank mRNA AK097741.1 - 2, 3 511
BC021728.2 - 2, 3 515
BI458986.1 - 2, 3 520

View Full Product Details