Gene Transcripts

Gene Symbol
ZNF75A
Entrez Gene ID
7627
Gene Name
zinc finger protein 75a
Gene Aliases
Chromosome Location
Chr. 16: 3305406 - 3322999 on Build GRCh38
UniGene ID
Hs.513292
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001302109.1 NP_001289038.1 6 1271
NM_001324041.1 NP_001310970.1 6 1228
NM_153028.3 NP_694573.1 5 747
NR_126069.1 4 906
NR_136681.1 5 1075
NR_136683.1 4 551
NR_136686.1 5 1101
XM_011522647.2 XP_011520949.1 5 845
XM_017023642.1 XP_016879131.1 5 643
GenBank mRNA AK056091.1 - 5 670
AK093979.1 - 4 859
BC119652.1 - 4 472 40115880
BC119653.1 - 4 472 40115881
CR749782.1 - 5 570

View Full Product Details