Gene Transcripts
- Gene Symbol
- KIF19
- Entrez Gene ID
- 124602
- Gene Name
- kinesin family member 19
- Gene Aliases
- KIF19A
- Chromosome Location
- Chr. 17: 74326212 - 74355820 on Build GRCh38
- UniGene ID
- Hs.372773
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_153209.3 |
NP_694941.2
|
4, 5 |
465 |
|
XM_005257029.1 |
XP_005257086.1
|
4, 5 |
465 |
|
XM_005257030.1 |
XP_005257087.1
|
4, 5 |
465 |
|
XM_017024150.1 |
XP_016879639.1
|
4, 5 |
465 |
|
XM_017024151.1 |
XP_016879640.1
|
4, 5 |
465 |
|
XM_017024152.1 |
XP_016879641.1
|
4, 5 |
465 |
|
XM_017024153.1 |
XP_016879642.1
|
4, 5 |
465 |
|
XM_017024154.1 |
XP_016879643.1
|
4, 5 |
465 |
|
XM_017024155.1 |
XP_016879644.1
|
4, 5 |
465 |
|
XM_017024156.1 |
XP_016879645.1
|
4, 5 |
465 |
|
XM_017024157.1 |
XP_016879646.1
|
4, 5 |
465 |
|
XM_017024158.1 |
XP_016879647.1
|
4, 5 |
465 |
|
GenBank mRNA |
AK094619.1 |
- |
4, 5 |
420 |
|
BC026362.1 |
- |
4, 5 |
446 |
|
BC093631.1 |
- |
4, 5 |
351 |
7939476 |
BC104852.1 |
- |
4, 5 |
351 |
8143855 |
BC110989.1 |
- |
4, 5 |
429 |
|
BC143327.1 |
- |
4, 5 |
352 |
9051832 |
DA955730.1 |
- |
4, 5 |
465 |
|
View Full Product Details