Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001160226.1 | NP_001153698.1 | 3 | 730 | |
NM_001160258.1 | NP_001153730.1 | 4 | 768 | ||
NM_001160259.1 | NP_001153731.1 | 2 | 643 | ||
NM_016083.4 | NP_057167.2 | 2 | 587 | ||
NM_033181.3 | NP_149421.2 | 2 | 242 | ||
XM_006715330.3 | XP_006715393.1 | 2 | 534 | ||
XM_011535424.1 | XP_011533726.1 | 3 | 1074 | ||
XM_011535425.1 | XP_011533727.1 | 4 | 1112 | ||
XM_011535426.1 | XP_011533728.1 | 5 | 1270 | ||
XM_011535428.1 | XP_011533730.1 | 3 | 921 | ||
XM_017010238.1 | XP_016865727.1 | 4 | 3898 | ||
XM_017010239.1 | XP_016865728.1 | 3 | 2221 | ||
XM_017010240.1 | XP_016865729.1 | 2 | 581 | ||
GenBank mRNA | AF107262.1 | - | 1 | 287 | |
AK313908.1 | - | 2 | 408 | ||
AY766182.1 | - | 2 | 179 | ||
AY995204.1 | - | 1 | 278 | ||
BC074811.2 | - | 1 | 316 | 30915316 | |
BC074812.2 | - | 1 | 316 | 30915532 | |
BC100968.1 | - | 1 | 278 | 40004970 | |
BC100969.1 | - | 1 | 278 | 40004975 | |
BC100970.1 | - | 1 | 278 | 40004978 | |
BC100971.1 | - | 1 | 278 | 40004979 | |
DQ067455.1 | - | 1 | 278 | ||
KC292264.1 | - | 2 | 242 | ||
X54937.1 | - | 2 | 426 | ||
X81120.1 | - | 1 | 369 | ||
X81121.1 | - | 1, 2 | 111 |