Gene Transcripts

Gene Symbol
AEBP2
Entrez Gene ID
121536
Gene Name
AE binding protein 2
Gene Aliases
Chromosome Location
Chr. 12: 19404101 - 19522239 on Build GRCh38
UniGene ID
Hs.126497
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114176.1 NP_001107648.1 9 2137
NM_001267043.1 NP_001253972.1 9 1587
NM_153207.4 NP_694939.2 8 2698
XM_006719031.3 XP_006719094.1 8 2205
XM_017018806.1 XP_016874295.1 9 1464
GenBank mRNA AB209384.1 - 9 1931
AK025788.1 - 1 1039
AK126973.1 - 9 2485
AK131361.1 - 9 2137
AK131410.1 - 9 1587
BC015624.2 - 8 2187 4843866
BC022220.1 - 8 2668 3914323

View Full Product Details