Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001200.3 | NP_001191.1 | 3 | 1246 | |
XM_011529323.2 | XP_011527625.1 | 2 | 1291 | ||
GenBank mRNA | BC069214.1 | - | 3 | 1246 | |
KC294426.1 | - | 2 | 461 | ||
M22489.1 | - | 3 | 784 |