Gene Transcripts

Gene Symbol
EYA2
Entrez Gene ID
2139
Gene Name
EYA transcriptional coactivator and phosphatase 2
Gene Aliases
EAB1
Chromosome Location
Chr. 20: 46894624 - 47188844 on Build GRCh38
UniGene ID
Hs.472877
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005244.4 NP_005235.3 6 818
NM_172110.3 NP_742108.2 6 818
XM_017027721.1 XP_016883210.1 6 536
GenBank mRNA AF055015.1 - 2 132
AK293032.1 - 6 522
AY705349.1 - 5 454
BC008803.2 - 6 500 3944547
BC013882.2 - 6 818
BT006682.1 - 5 372
U71207.1 - 5 446

View Full Product Details