Gene Transcripts

Gene Symbol
FNDC7
Entrez Gene ID
163479
Gene Name
fibronectin type III domain containing 7
Gene Aliases
Chromosome Location
Chr. 1: 108712934 - 108742749 on Build GRCh38
UniGene ID
Hs.258253
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144937.1 NP_001138409.1 10 2109
GenBank mRNA AK093157.1 - 6 1435
BC057762.1 - 6 1482
BC130421.1 - 6 1435 40146452

View Full Product Details