Gene Transcripts

Gene Symbol
SLC39A5
Entrez Gene ID
283375
Gene Name
solute carrier family 39 member 5
Gene Aliases
LZT-Hs7, MYP24, ZIP5
Chromosome Location
Chr. 12: 56230036 - 56237846 on Build GRCh38
UniGene ID
Hs.591018
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135195.1 NP_001128667.1 7 1204
NM_173596.2 NP_775867.2 9 1233
XM_005268803.1 XP_005268860.1 7 1121
XM_011538198.1 XP_011536500.1 7 1186
XM_011538199.1 XP_011536501.1 9 1240
XM_011538200.1 XP_011536502.1 8 1190
XM_011538201.1 XP_011536503.1 8 1164
XM_017019185.1 XP_016874674.1 7 1164
XM_017019186.1 XP_016874675.1 9 1373
XM_017019187.1 XP_016874676.1 7 1122
GenBank mRNA AK172768.1 - 7 1167
AK313188.1 - 9 1196
BC027884.1 - 7 1078 5221329

View Full Product Details