Gene Transcripts
- Gene Symbol
- RSPO1
- Entrez Gene ID
- 284654
- Gene Name
- R-spondin 1
- Gene Aliases
- CRISTIN3, RSPO
- Chromosome Location
- Chr. 1: 37611350 - 37634923 on Build GRCh38
- UniGene ID
- Hs.135015
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK098225.1 |
- |
7 |
2220 |
|
DQ165084.1 |
- |
7 |
2541 |
|
DQ165085.1 |
- |
6 |
2352 |
|
View Full Product Details