Gene Transcripts

Gene Symbol
NKX2-2
Entrez Gene ID
4821
Gene Name
NK2 homeobox 2
Gene Aliases
NKX2.2, NKX2B
Chromosome Location
Chr. 20: 21511010 - 21514026 on Build GRCh38
UniGene ID
Hs.516922
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002509.3 NP_002500.1 2 695
XM_006723566.3 XP_006723629.1 2 352
GenBank mRNA BC075092.2 - 2 366
BC075093.2 - 2 366 30915391

View Full Product Details