Gene Transcripts
- Gene Symbol
- HSPB1
- Entrez Gene ID
- 3315
- Gene Name
- heat shock protein family B (small) member 1
- Gene Aliases
- CMT2F, HEL-S-102, HMN2B, HS.76067, HSP27, HSP28, Hsp25, SRP27
- Chromosome Location
- Chr. 7: 76302558 - 76304297 on Build GRCh38
- UniGene ID
- Hs.520973
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
FJ224323.1 |
- |
3 |
730 |
|
U90906.1 |
- |
3 |
730 |
|
View Full Product Details