Gene Transcripts

Gene Symbol
AQP5
Entrez Gene ID
362
Gene Name
aquaporin 5
Gene Aliases
AQP-5, PPKB
Chromosome Location
Chr. 12: 49961496 - 49965682 on Build GRCh38
UniGene ID
Hs.298023
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001651.3 NP_001642.1 4 1232
GenBank mRNA BC032946.1 - 4 782
BP272197.1 - 2 119
CR541989.1 - 4 710
CR542022.1 - 4 710

View Full Product Details