Gene Transcripts

Gene Symbol
SMAD2
Entrez Gene ID
4087
Gene Name
SMAD family member 2
Gene Aliases
JV18, JV18-1, MADH2, MADR2, hMAD-2, hSMAD2
Chromosome Location
Chr. 18: 47833095 - 47931193 on Build GRCh38
UniGene ID
Hs.12253
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003652.3 NP_001003652.1 11 1828
NM_001135937.2 NP_001129409.1 10 1738
NM_005901.5 NP_005892.1 11 1720
XM_005258259.3 XP_005258316.1 12 1961
XM_006722451.3 XP_006722514.1 12 1973
XM_011525984.2 XP_011524286.1 11 1317
XM_011525985.2 XP_011524287.1 13 2020
XM_017025745.1 XP_016881234.1 13 2059
XM_017025746.1 XP_016881235.1 11 1871
XM_017025747.1 XP_016881236.1 12 1946
XM_017025748.1 XP_016881237.1 13 2032
XM_017025750.1 XP_016881239.1 12 1921
GenBank mRNA AF027964.1 - 11 1534
BC014840.1 - 11 1676 4449540
BC025699.1 - 11 1534 5221801

View Full Product Details