Gene Transcripts

Gene Symbol
ROMO1
Entrez Gene ID
140823
Gene Name
reactive oxygen species modulator 1
Gene Aliases
C20orf52, MTGM, MTGMP, bA353C18.2
Chromosome Location
Chr. 20: 35699310 - 35700982 on Build GRCh38
UniGene ID
Hs.472564
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080748.2 NP_542786.1 3 437
XM_017027678.1 XP_016883167.1 3 432
GenBank mRNA AK000548.1 - 3 342
BC008488.1 - 453 4133274

View Full Product Details