Gene Transcripts

Gene Symbol
METTL26
Entrez Gene ID
84326
Gene Name
methyltransferase like 26
Gene Aliases
C16orf13, JFP2
Chromosome Location
Chr. 16: 634427 - 636366 on Build GRCh38
UniGene ID
Hs.239500
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040160.2 NP_001035250.1 5 775
NM_001040161.2 NP_001035251.1 3 552
NM_001040162.2 NP_001035252.1 4 612
NM_001040165.2 NP_001035255.1 4 715
NM_001288710.1 NP_001275639.1 5 783
NM_032366.4 NP_115742.3 6 843
NR_109976.1 5 787
NR_109977.1 5 680
NR_109978.1 6 950
NR_109979.1 4 620
XM_011522713.2 XP_011521015.1 6 1042
XM_011522714.2 XP_011521016.1 5 867
GenBank mRNA AK057077.1 - 3 1741
AK294482.1 - 6 779
AK295628.1 - 5 711
BC007207.1 - 3 534 4054799
BU539308.1 - 4 568
C00865.1 - 1 171

View Full Product Details