Gene Transcripts
- Gene Symbol
- CDH26
- Entrez Gene ID
- 60437
- Gene Name
- cadherin 26
- Gene Aliases
- VR20
- Chromosome Location
- Chr. 20: 59957764 - 60035629 on Build GRCh38
- UniGene ID
- Hs.729046
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021810.4 |
NP_068582.2
|
2, 3 |
304 |
|
NM_177980.2 |
NP_817089.1
|
14, 15 |
2407 |
|
XM_011528969.2 |
XP_011527271.1
|
14, 15 |
2413 |
|
XM_011528970.2 |
XP_011527272.1
|
8, 9 |
1429 |
|
XM_017027994.1 |
XP_016883483.1
|
14, 15 |
3076 |
|
XM_017027996.1 |
XP_016883485.1
|
14, 15 |
2978 |
|
XM_017027998.1 |
XP_016883487.1
|
14, 15 |
3074 |
|
XM_017028000.1 |
XP_016883489.1
|
13, 14 |
2246 |
|
XM_017028002.1 |
XP_016883491.1
|
14, 15 |
3075 |
|
GenBank mRNA |
AK055202.1 |
- |
14, 15 |
2396 |
|
AK074477.1 |
- |
2, 3 |
292 |
|
BC136306.1 |
- |
14, 15 |
2211 |
9020293 |
BC143931.1 |
- |
14, 15 |
2211 |
|
View Full Product Details