Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030876.5 | NP_110503.3 | 1 | 482 | |
XM_011514929.2 | XP_011513231.1 | 3 | 657 | ||
XM_011514930.2 | XP_011513232.1 | 2 | 535 | ||
GenBank mRNA | AJ459859.1 | - | 1 | 485 | |
BC069149.1 | - | 1 | 400 |