Gene Transcripts

Gene Symbol
VN1R2
Entrez Gene ID
317701
Gene Name
vomeronasal 1 receptor 2
Gene Aliases
V1RL2
Chromosome Location
Chr. 19: 53258292 - 53259602 on Build GRCh38
UniGene ID
Hs.553684
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173856.2 NP_776255.2 1 457
GenBank mRNA BC130356.1 - 1 457
BC130358.1 - 1 457 40146389

View Full Product Details