Gene Transcripts

Gene Symbol
MYRF
Entrez Gene ID
745
Gene Name
myelin regulatory factor
Gene Aliases
C11orf9, MRF, Ndt80, pqn-47
Chromosome Location
Chr. 11: 61752617 - 61788518 on Build GRCh38
UniGene ID
Hs.473109
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127392.2 NP_001120864.1 2 208
NM_013279.3 NP_037411.1 2 108
XM_005274222.1 XP_005274279.1 2 196
XM_005274223.1 XP_005274280.1 2 196
XM_005274224.1 XP_005274281.1 2 196
XM_005274225.1 XP_005274282.1 2 196
XM_005274226.1 XP_005274283.1 2 196
XM_005274227.1 XP_005274284.1 2 196
XM_005274228.1 XP_005274285.1 2 196
XM_011545234.2 XP_011543536.1 2 228
GenBank mRNA AB023171.2 - 2 196
AF086762.1 - 2 108
HY145048.1 - 2 210
HY148147.1 - 2 208

View Full Product Details