Gene Transcripts
- Gene Symbol
- CBWD1
- Entrez Gene ID
- 55871
- Gene Name
- COBW domain containing 1
- Gene Aliases
- COBP
- Chromosome Location
- Chr. 9: 121038 - 179083 on Build GRCh38
- UniGene ID
- Hs.531330
- Species
- Homo sapiens
- Gene Symbol
- CBWD2
- Entrez Gene ID
- 150472
- Gene Name
- COBW domain containing 2
- Gene Aliases
- Chromosome Location
- Chr. 2: 113437691 - 113496204 on Build GRCh38
- UniGene ID
- Hs.531330
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK225251.1 |
- |
16 |
1349 |
|
View Full Product Details