Gene Transcripts

Gene Symbol
DISP1
Entrez Gene ID
84976
Gene Name
dispatched RND transporter family member 1
Gene Aliases
DISPA
Chromosome Location
Chr. 1: 222814514 - 223005995 on Build GRCh38
UniGene ID
Hs.528817
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032890.3 NP_116279.2 4 451
XM_005273335.2 XP_005273392.1 2 292
XM_006711592.2 XP_006711655.1 3 1239
XM_011510072.2 XP_011508374.1 3 291
XM_011510073.2 XP_011508375.1 3 769
XM_011510074.2 XP_011508376.1 3 310
XM_011510075.2 XP_011508377.1 3 797
XM_011510077.2 XP_011508379.1 2 233
XM_017002611.1 XP_016858100.1 2 707
GenBank mRNA AK098669.1 - 2 185
AK125728.1 - 2 289
BC007734.2 - 2 272
BC107762.1 - 2 276
DA756409.1 - 4 451

View Full Product Details