Gene Transcripts
- Gene Symbol
- NUP210P1
- Entrez Gene ID
- 255330
- Gene Name
- nucleoporin 210 pseudogene 1
- Gene Aliases
- C3orf46
- Chromosome Location
- Chr. 3: 126662081 - 126671939 on Build GRCh38
- UniGene ID
- Hs.145435
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_034158.1 |
|
4, 5 |
637 |
|
GenBank mRNA |
BC042038.1 |
- |
4, 5 |
637 |
|
View Full Product Details