Gene Transcripts

Gene Symbol
CLK2
Entrez Gene ID
1196
Gene Name
CDC like kinase 2
Gene Aliases
Chromosome Location
Chr. 1: 155262868 - 155273529 on Build GRCh38
UniGene ID
Hs.73986
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001294338.1 NP_001281267.1 12 1567
NM_001294339.1 NP_001281268.1 11 1479
NM_003993.3 NP_003984.2 12 1564
XM_005244878.1 XP_005244935.1 12 1546
XM_011509143.1 XP_011507445.1 12 1549
XM_011509147.2 XP_011507449.1 11 1444
XM_017000241.1 XP_016855730.1 11 1447
XM_017000242.1 XP_016855731.1 10 1359
XM_017000243.1 XP_016855732.1 9 2487
XM_017000244.1 XP_016855733.1 11 1441
GenBank mRNA AK091036.1 - 6 2102
AK291995.1 - 12 1411
AK303696.1 - 12 1258
BC006274.2 - 8 755
BC014067.2 - 12 1322 3837777
BC053603.1 - 12 1485 6007375
L29216.1 - 11 1279
L29218.1 - 12 1367

View Full Product Details